Methyl 2-[(2,5-dichlorophenyl)formamido]acetate, tech
Catalog # APOSOR32988-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:262.089 g/mol
Storage Temperature:Ambient
CAS Number:338965-44-9
Formula:C₁₀H₉Cl₂NO₃
MDL Number:MFCD01195627