Ethyl 4-hydroxy-6,8-dimethylquinoline-3-carboxylate 95%
Catalog # APOSOR32979-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:245.274 g/mol
Storage Temperature:Ambient
CAS Number:77156-77-5
Formula:C₁₄H₁₅NO₃
MDL Number:MFCD00173389