Order Entry
Czech Republic
ContactUsLinkComponent
 
(13S,17R)-16-Methyl-11,15-dioxa-16-azatetracyclo[8.7.0.02,7.013,17]heptadeca-1(10),2(7),3,5,8-pentaene, tech
Catalog # APOSOR32968-1G
Supplier:  Apollo Scientific
CAS Number:  
(13S,17R)-16-Methyl-11,15-dioxa-16-azatetracyclo[8.7.0.02,7.013,17]heptadeca-1(10),2(7),3,5,8-pentaene, tech
Catalog # APOSOR32968-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :241.285 g/mol
Storage Temperature:Ambient
CAS Number:28082-15-7
Formula:C₁₅H₁₅NO₂
MDL Number:MFCD00231681