5-Chloro-6-methyl-2-phenylpyrimidin-4-ol 95%
Catalog # APOSOR32952-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:220.655 g/mol
Storage Temperature:Ambient
CAS Number:20551-31-9
Formula:C₁₁H₉ClN₂O
MDL Number:MFCD00232127