Order Entry
Czech Republic
ContactUsLinkComponent
2,2-Dimethyl-5-[(4-methylphenyl)methylidene]-1,3-dioxane-4,6-dione, tech
2,2-Dimethyl-5-[(4-methylphenyl)methylidene]-1,3-dioxane-4,6-dione, tech
Catalog # APOSOR32949-1G
Supplier:  Apollo Scientific
CAS Number:  
2,2-Dimethyl-5-[(4-methylphenyl)methylidene]-1,3-dioxane-4,6-dione, tech
Catalog # APOSOR32949-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :246.259 g/mol
Storage Temperature:Ambient
CAS Number:15795-51-4
Formula:C₁₄H₁₄O₄
MDL Number:MFCD00128563