Order Entry
Czech Republic
ContactUsLinkComponent
 
4-[4-(1H-Pyrrol-1-yl)phenyl]-1H-pyrazol-5-amine, tech
Catalog # APOSOR32936-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[4-(1H-Pyrrol-1-yl)phenyl]-1H-pyrazol-5-amine, tech
Catalog # APOSOR32936-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :224.261 g/mol
Storage Temperature:Ambient
CAS Number:1024019-01-9
Formula:C₁₃H₁₂N₄
MDL Number:MFCD04125700