3-{6-Chloroimidazo[1,2-a]pyridin-2-yl}aniline, tech
Catalog # APOSOR32917-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.692 g/mol
Storage Temperature:Ambient
CAS Number:439110-86-8
Formula:C₁₃H₁₀ClN₃
MDL Number:MFCD03012831