(2E)-3-(Furan-2-yl)-1-(pyridin-3-yl)prop-2-en-1-one, tech
Catalog # APOSOR32880-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:1 g
- Sekvence:3047
Specifications
About this item
:
:199.205 g/mol
Storage Temperature:Ambient
CAS Number:18461-28-4
Formula:C₁₂H₉NO₂
MDL Number:MFCD00224733