
4-Aminopyrimidine hydrochloride
Catalog # APOSOR3288-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:131.564 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:174500-31-3
Formula:C₄H₅N₃·ClH
MDL Number:MFCD09702435