Order Entry
Czech Republic
ContactUsLinkComponent
(2E)-1,3-Bis(furan-2-yl)prop-2-en-1-one, tech
(2E)-1,3-Bis(furan-2-yl)prop-2-en-1-one, tech
Catalog # APOSOR32879-1G
Supplier:  Apollo Scientific
CAS Number:  
(2E)-1,3-Bis(furan-2-yl)prop-2-en-1-one, tech
Catalog # APOSOR32879-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :188.179 g/mol
Storage Temperature:Ambient
CAS Number:42811-85-8
Formula:C₁₁H₈O₃
MDL Number:MFCD00020927