Order Entry
Czech Republic
ContactUsLinkComponent
(2E)-3-(Furan-2-yl)-1-(pyridin-2-yl)prop-2-en-1-one, tech
(2E)-3-(Furan-2-yl)-1-(pyridin-2-yl)prop-2-en-1-one, tech
Catalog # APOSOR32878-1G
Supplier:  Apollo Scientific
CAS Number:  
(2E)-3-(Furan-2-yl)-1-(pyridin-2-yl)prop-2-en-1-one, tech
Catalog # APOSOR32878-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :199.205 g/mol
Storage Temperature:Ambient
CAS Number:18551-00-3
Formula:C₁₂H₉NO₂
MDL Number:MFCD01315211