Order Entry
Czech Republic
ContactUsLinkComponent
 
(3E)-3-{[(2,4-Dichlorophenyl)methoxy]imino}propanenitrile, tech
Catalog # APOSOR32865-1G
Supplier:  Apollo Scientific
CAS Number:  
(3E)-3-{[(2,4-Dichlorophenyl)methoxy]imino}propanenitrile, tech
Catalog # APOSOR32865-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :243.089 g/mol
Storage Temperature:Ambient
CAS Number:338959-93-6
Formula:C₁₀H₈Cl₂N₂O
MDL Number:MFCD00663727