Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 4-hydroxy-5,7-dimethylquinoline-3-carboxylate, tech
Ethyl 4-hydroxy-5,7-dimethylquinoline-3-carboxylate, tech
Catalog # APOSOR32852-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 4-hydroxy-5,7-dimethylquinoline-3-carboxylate, tech
Catalog # APOSOR32852-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :245.274 g/mol
Storage Temperature:Ambient
CAS Number:93514-84-2
Formula:C₁₄H₁₅NO₃
MDL Number:MFCD00173347