Order Entry
Czech Republic
ContactUsLinkComponent
1-(4,6-Dimethylpyridin-2-yl)-3-methylurea, tech
1-(4,6-Dimethylpyridin-2-yl)-3-methylurea, tech
Catalog # APOSOR32850-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4,6-Dimethylpyridin-2-yl)-3-methylurea, tech
Catalog # APOSOR32850-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :179.219 g/mol
Storage Temperature:Ambient
CAS Number:866038-54-2
Formula:C₉H₁₃N₃O
MDL Number:MFCD04125586