Order Entry
Czech Republic
ContactUsLinkComponent
1,3-Diphenyl-4,5-dihydro-1H-pyrazol-5-one 95%
1,3-Diphenyl-4,5-dihydro-1H-pyrazol-5-one 95%
Catalog # APOSOR32837-1G
Supplier:  Apollo Scientific
CAS Number:  
1,3-Diphenyl-4,5-dihydro-1H-pyrazol-5-one 95%
Catalog # APOSOR32837-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :236.269 g/mol
Storage Temperature:Ambient
CAS Number:4845-49-2
Formula:C₁₅H₁₂N₂O
MDL Number:MFCD00117375