Order Entry
Czech Republic
ContactUsLinkComponent
2,2-Dimethyl-5-[(2E)-3-phenylprop-2-en-1-ylidene]-1,3-dioxane-4,6-dione, tech
2,2-Dimethyl-5-[(2E)-3-phenylprop-2-en-1-ylidene]-1,3-dioxane-4,6-dione, tech
Catalog # APOSOR32812-1G
Supplier:  Apollo Scientific
CAS Number:  
2,2-Dimethyl-5-[(2E)-3-phenylprop-2-en-1-ylidene]-1,3-dioxane-4,6-dione, tech
Catalog # APOSOR32812-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :258.269 g/mol
Storage Temperature:Ambient
CAS Number:548765-13-5
Formula:C₁₅H₁₄O₄
MDL Number:MFCD00128548