5-(Prop-2-yn-1-yloxy)-2H-1,3-benzodioxole, tech
Catalog # APOSOR32791-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:176.169 g/mol
Storage Temperature:Ambient
CAS Number:19947-80-9
Formula:C₁₀H₈O₃
MDL Number:MFCD03787731