Order Entry
Czech Republic
ContactUsLinkComponent
1-Methyl-4-(3-phenylprop-2-yn-1-yl)piperazine, tech
1-Methyl-4-(3-phenylprop-2-yn-1-yl)piperazine, tech
Catalog # APOSOR32790-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Methyl-4-(3-phenylprop-2-yn-1-yl)piperazine, tech
Catalog # APOSOR32790-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :214.306 g/mol
Storage Temperature:Ambient
CAS Number:13808-26-9
Formula:C₁₄H₁₈N₂
MDL Number:MFCD03787141