Order Entry
Czech Republic
ContactUsLinkComponent
6-(Methoxymethyl)-N-methyl-2-phenylpyrimidin-4-amine, tech
6-(Methoxymethyl)-N-methyl-2-phenylpyrimidin-4-amine, tech
Catalog # APOSOR32762-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(Methoxymethyl)-N-methyl-2-phenylpyrimidin-4-amine, tech
Catalog # APOSOR32762-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :229.278 g/mol
Storage Temperature:Ambient
CAS Number:339278-91-0
Formula:C₁₃H₁₅N₃O
MDL Number:MFCD00794771