Order Entry
Czech Republic
ContactUsLinkComponent
3-(4-Methoxyphenyl)-1-[(E)-2-phenylethenyl]urea, tech
3-(4-Methoxyphenyl)-1-[(E)-2-phenylethenyl]urea, tech
Catalog # APOSOR32756-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Methoxyphenyl)-1-[(E)-2-phenylethenyl]urea, tech
Catalog # APOSOR32756-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :268.31 g/mol
Storage Temperature:Ambient
CAS Number:74088-06-5
Formula:C₁₆H₁₆N₂O₂
MDL Number:MFCD00794891