2-Methyl-3-phenyl-octahydro-1,2-benzoxazol-4-one, tech
Catalog # APOSOR32752-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:231.29 g/mol
Storage Temperature:Ambient
CAS Number:318248-52-1
Formula:C₁₄H₁₇NO₂
MDL Number:MFCD00974227