Order Entry
Czech Republic
ContactUsLinkComponent
5-(4-Chlorophenyl)-2-oxo-1-(propan-2-yl)-1,2-dihydropyridine-3-carbonitrile, tech
5-(4-Chlorophenyl)-2-oxo-1-(propan-2-yl)-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSOR32745-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(4-Chlorophenyl)-2-oxo-1-(propan-2-yl)-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSOR32745-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :272.73 g/mol
Storage Temperature:Ambient
CAS Number:338423-60-2
Formula:C₁₅H₁₃ClN₂O
MDL Number:MFCD00231550