Order Entry
Czech Republic
ContactUsLinkComponent
2-(3-Methoxyphenyl)-2-(morpholin-4-yl)acetonitrile 95%
2-(3-Methoxyphenyl)-2-(morpholin-4-yl)acetonitrile 95%
Catalog # APOSOR32733-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(3-Methoxyphenyl)-2-(morpholin-4-yl)acetonitrile 95%
Catalog # APOSOR32733-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :232.278 g/mol
Storage Temperature:Ambient
CAS Number:123567-57-7
Formula:C₁₃H₁₆N₂O₂
MDL Number:MFCD00172615