Order Entry
Czech Republic
ContactUsLinkComponent
1-(2,3-Dihydro-1,4-benzodioxin-6-yl)-3-(2-methylprop-1-en-1-yl)urea, tech
1-(2,3-Dihydro-1,4-benzodioxin-6-yl)-3-(2-methylprop-1-en-1-yl)urea, tech
Catalog # APOSOR32698-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2,3-Dihydro-1,4-benzodioxin-6-yl)-3-(2-methylprop-1-en-1-yl)urea, tech
Catalog # APOSOR32698-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :248.278 g/mol
Storage Temperature:Ambient
CAS Number:865659-94-5
Formula:C₁₃H₁₆N₂O₃
MDL Number:MFCD04125278