2-Phenylimidazo[1,2-a]pyridine-6-carbonitrile, tech
Catalog # APOSOR32682-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.241 g/mol
Storage Temperature:Ambient
CAS Number:214958-29-9
Formula:C₁₄H₉N₃
MDL Number:MFCD03787072