Order Entry
Czech Republic
ContactUsLinkComponent
6-(4-Chlorobenzenesulphonyl)-1-oxa-6-azaspiro[2.5]octane, tech
6-(4-Chlorobenzenesulphonyl)-1-oxa-6-azaspiro[2.5]octane, tech
Catalog # APOSOR32666-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(4-Chlorobenzenesulphonyl)-1-oxa-6-azaspiro[2.5]octane, tech
Catalog # APOSOR32666-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :287.762 g/mol
Storage Temperature:Ambient
CAS Number:861212-71-7
Formula:C₁₂H₁₄ClNO₃S
MDL Number:MFCD04125157