Order Entry
Czech Republic
ContactUsLinkComponent
2-(Phenylamino)-2,3,3a,4,7,7a-hexahydro-1H-isoindole-1,3-dione, tech
2-(Phenylamino)-2,3,3a,4,7,7a-hexahydro-1H-isoindole-1,3-dione, tech
Katalogové číslo APOSOR32661-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Phenylamino)-2,3,3a,4,7,7a-hexahydro-1H-isoindole-1,3-dione, tech
Katalogové číslo APOSOR32661-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :242.273 g/mol
Storage Temperature:Ambient
CAS Number:61152-63-4
Formula:C₁₄H₁₄N₂O₂
MDL Number:MFCD01717970