Order Entry
Czech Republic
ContactUsLinkComponent
 
(2E)-3-(4-Methoxyphenyl)-1-(pyridin-2-yl)prop-2-en-1-one, tech
Catalog # APOSOR32649-1G
Supplier:  Apollo Scientific
CAS Number:  
(2E)-3-(4-Methoxyphenyl)-1-(pyridin-2-yl)prop-2-en-1-one, tech
Catalog # APOSOR32649-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :239.269 g/mol
Storage Temperature:Ambient
CAS Number:18451-44-0
Formula:C₁₅H₁₃NO₂
MDL Number:MFCD01315417