Order Entry
Czech Republic
ContactUsLinkComponent
3-(Dimethylamino)-1-(3-methylthiophen-2-yl)but-2-en-1-one, tech
3-(Dimethylamino)-1-(3-methylthiophen-2-yl)but-2-en-1-one, tech
Catalog # APOSOR32576-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Dimethylamino)-1-(3-methylthiophen-2-yl)but-2-en-1-one, tech
Catalog # APOSOR32576-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :209.308 g/mol
Storage Temperature:Ambient
CAS Number:861209-49-6
Formula:C₁₁H₁₅NOS
MDL Number:MFCD03618011