Order Entry
Czech Republic
ContactUsLinkComponent
4-[(4-Chlorophenyl)methoxy]-1lambda6-thiomorpholine-1,1-dione, tech
4-[(4-Chlorophenyl)methoxy]-1lambda6-thiomorpholine-1,1-dione, tech
Catalog # APOSOR32567-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[(4-Chlorophenyl)methoxy]-1lambda6-thiomorpholine-1,1-dione, tech
Catalog # APOSOR32567-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :275.752 g/mol
Storage Temperature:Ambient
CAS Number:477889-69-3
Formula:C₁₁H₁₄ClNO₃S
MDL Number:MFCD02102601