Order Entry
Czech Republic
ContactUsLinkComponent
5-(4-Chlorophenyl)-2-[(dimethylamino)methylidene]cyclohexane-1,3-dione, tech
5-(4-Chlorophenyl)-2-[(dimethylamino)methylidene]cyclohexane-1,3-dione, tech
Katalogové číslo APOSOR32536-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(4-Chlorophenyl)-2-[(dimethylamino)methylidene]cyclohexane-1,3-dione, tech
Katalogové číslo APOSOR32536-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :277.746 g/mol
Storage Temperature:Ambient
CAS Number:338399-40-9
Formula:C₁₅H₁₆ClNO₂
MDL Number:MFCD00172130