Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl (2E)-3-amino-3-(4-phenylpiperazin-1-yl)prop-2-enoate, tech
Ethyl (2E)-3-amino-3-(4-phenylpiperazin-1-yl)prop-2-enoate, tech
Catalog # APOSOR32529-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl (2E)-3-amino-3-(4-phenylpiperazin-1-yl)prop-2-enoate, tech
Catalog # APOSOR32529-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :275.346 g/mol
Storage Temperature:Ambient
CAS Number:79823-34-0
Formula:C₁₅H₂₁N₃O₂
MDL Number:MFCD00664733