Order Entry
Czech Republic
ContactUsLinkComponent
1-Acetyl-N-(3-ethynylphenyl)piperidine-4-carboxamide, tech
1-Acetyl-N-(3-ethynylphenyl)piperidine-4-carboxamide, tech
Catalog # APOSOR32514-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Acetyl-N-(3-ethynylphenyl)piperidine-4-carboxamide, tech
Catalog # APOSOR32514-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :270.326 g/mol
Storage Temperature:Ambient
CAS Number:861208-20-0
Formula:C₁₆H₁₈N₂O₂
MDL Number:MFCD06496153