Order Entry
Czech Republic
ContactUsLinkComponent
(E)-N-[(3-Chloro-4-methoxyphenyl)methylidene]hydroxylamine 95%
(E)-N-[(3-Chloro-4-methoxyphenyl)methylidene]hydroxylamine 95%
Catalog # APOSOR32506-1G
Supplier:  Apollo Scientific
CAS Number:  
(E)-N-[(3-Chloro-4-methoxyphenyl)methylidene]hydroxylamine 95%
Catalog # APOSOR32506-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :185.608 g/mol
Storage Temperature:Ambient
CAS Number:861207-68-3
Formula:C₈H₈ClNO₂
MDL Number:MFCD04124816