Order Entry
Czech Republic
ContactUsLinkComponent
Cyanomethyl 4-(1H-pyrrol-1-yl)benzoate, tech
Cyanomethyl 4-(1H-pyrrol-1-yl)benzoate, tech
Catalog # APOSOR32481-1G
Supplier:  Apollo Scientific
CAS Number:  
Cyanomethyl 4-(1H-pyrrol-1-yl)benzoate, tech
Catalog # APOSOR32481-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :226.231 g/mol
Storage Temperature:Ambient
CAS Number:439095-55-3
Formula:C₁₃H₁₀N₂O₂
MDL Number:MFCD03012862