(2E)-1-(5-Chlorothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1276536-34-5
:
Formula:C₉H₁₀ClNOS
MDL Number:MFCD03617978
:215.7 g/mol
Storage Temperature:Ambient
Frequently Bought Together