Order Entry
Czech Republic
ContactUsLinkComponent
(2E)-1-(1-Benzothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one, tech
(2E)-1-(1-Benzothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one, tech
Catalog # APOSOR32471-1G
Supplier:  Apollo Scientific
CAS Number:  
(2E)-1-(1-Benzothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one, tech
Catalog # APOSOR32471-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :231.313 g/mol
Storage Temperature:Ambient
CAS Number:893441-59-3
Formula:C₁₃H₁₃NOS
MDL Number:MFCD03617977