Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(2,4-Dichlorophenyl)-3-hydroxy-3-methylurea
1-(2,4-Dichlorophenyl)-3-hydroxy-3-methylurea, tech
1-(2,4-Dichlorophenyl)-3-hydroxy-3-methylurea, tech
APOSOR32446-1G
:
Apollo Scientific
:
1-(2,4-Dichlorophenyl)-3-hydroxy-3-methylurea, tech
APOSOR32446-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
235.067 g/mol
:
Ambient
:
28267-53-0
:
C₈H₈Cl₂N₂O₂
:
MFCD02082386