Order Entry
Czech Republic
ContactUsLinkComponent
{7-Methyl-2-phenylimidazo[1,2-a]pyridin-3-yl}methanol, tech
{7-Methyl-2-phenylimidazo[1,2-a]pyridin-3-yl}methanol, tech
Catalog # APOSOR32440-1G
Supplier:  Apollo Scientific
CAS Number:  
{7-Methyl-2-phenylimidazo[1,2-a]pyridin-3-yl}methanol, tech
Catalog # APOSOR32440-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :238.284 g/mol
Storage Temperature:Ambient
CAS Number:217435-73-9
Formula:C₁₅H₁₄N₂O
MDL Number:MFCD01815092