Order Entry
Czech Republic
ContactUsLinkComponent
[4-(4-Methoxyphenoxy)phenyl]methanamine 95%
[4-(4-Methoxyphenoxy)phenyl]methanamine 95%
Catalog # APOSOR32439-1G
Supplier:  Apollo Scientific
CAS Number:  
[4-(4-Methoxyphenoxy)phenyl]methanamine 95%
Catalog # APOSOR32439-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :229.274 g/mol
Storage Temperature:Ambient
CAS Number:477868-65-8
Formula:C₁₄H₁₅NO₂
MDL Number:MFCD01814768