Order Entry
Czech Republic
ContactUsLinkComponent
3-[(2-Aminophenyl)amino]-5,5-dimethylcyclohex-2-en-1-one, tech
3-[(2-Aminophenyl)amino]-5,5-dimethylcyclohex-2-en-1-one, tech
Catalog # APOSOR32429-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[(2-Aminophenyl)amino]-5,5-dimethylcyclohex-2-en-1-one, tech
Catalog # APOSOR32429-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :230.306 g/mol
Storage Temperature:Ambient
CAS Number:39222-69-0
Formula:C₁₄H₁₈N₂O
MDL Number:MFCD00156635