Order Entry
Czech Republic
ContactUsLinkComponent
4-Chloro-2-(4-methoxyphenyl)-5-[(prop-2-en-1-yl)amino]-2,3-dihydropyridazin-3-one, tech
4-Chloro-2-(4-methoxyphenyl)-5-[(prop-2-en-1-yl)amino]-2,3-dihydropyridazin-3-one, tech
Catalog # APOSOR32417-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-2-(4-methoxyphenyl)-5-[(prop-2-en-1-yl)amino]-2,3-dihydropyridazin-3-one, tech
Catalog # APOSOR32417-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :291.733 g/mol
Storage Temperature:Ambient
CAS Number:477867-47-3
Formula:C₁₄H₁₄ClN₃O₂
MDL Number:MFCD00793839