Order Entry
Czech Republic
ContactUsLinkComponent
(2E)-1-(3-Chlorothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one, tech
(2E)-1-(3-Chlorothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one, tech
Catalog # APOSOR32407-1G
Supplier:  Apollo Scientific
CAS Number:  
(2E)-1-(3-Chlorothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one, tech
Catalog # APOSOR32407-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :215.7 g/mol
Storage Temperature:Ambient
CAS Number:166196-77-6
Formula:C₉H₁₀ClNOS
MDL Number:MFCD00793820