Order Entry
Czech Republic
ContactUsLinkComponent
O-[(2,5-Dichlorophenyl)methyl]hydroxylamine hydrochloride 95%
O-[(2,5-Dichlorophenyl)methyl]hydroxylamine hydrochloride 95%
Catalog # APOSOR32398-1G
Supplier:  Apollo Scientific
CAS Number:  
O-[(2,5-Dichlorophenyl)methyl]hydroxylamine hydrochloride 95%
Catalog # APOSOR32398-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :228.504 g/mol
Storage Temperature:Ambient
CAS Number:317821-71-9
Formula:C₇H₇Cl₂NO·HCl
MDL Number:MFCD01114576