7-(4-Bromophenyl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech
Catalog # APOSOR32382-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:299.125 g/mol
Storage Temperature:Ambient
CAS Number:320417-25-2
Formula:C₁₃H₇BrN₄
MDL Number:MFCD00138886