Order Entry
Czech Republic
ContactUsLinkComponent
1-[(1-Phenyl-1H-1,2,4-triazol-3-yl)oxy]propan-2-one, tech
1-[(1-Phenyl-1H-1,2,4-triazol-3-yl)oxy]propan-2-one, tech
Katalogové číslo APOSOR32377-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[(1-Phenyl-1H-1,2,4-triazol-3-yl)oxy]propan-2-one, tech
Katalogové číslo APOSOR32377-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :217.224 g/mol
Storage Temperature:Ambient
CAS Number:320416-23-7
Formula:C₁₁H₁₁N₃O₂
MDL Number:MFCD00138721