Order Entry
Czech Republic
ContactUsLinkComponent
(4Z)-4-[(Dimethylamino)methylidene]-3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one 95%
(4Z)-4-[(Dimethylamino)methylidene]-3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one 95%
Catalog # APOSOR32370-1G
Supplier:  Apollo Scientific
CAS Number:  
(4Z)-4-[(Dimethylamino)methylidene]-3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one 95%
Catalog # APOSOR32370-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :229.278 g/mol
Storage Temperature:Ambient
CAS Number:65644-13-5
Formula:C₁₃H₁₅N₃O
MDL Number:MFCD00100702