1-[2-(1H-Imidazol-1-yl)phenyl]ethan-1-one, tech
Catalog # APOSOR32368-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:186.21 g/mol
Storage Temperature:Ambient
CAS Number:20513-61-5
Formula:C₁₁H₁₀N₂O
MDL Number:MFCD00127221