Order Entry
Czech Republic
ContactUsLinkComponent
1,2,3-Trimethoxy-5-(prop-2-yn-1-yloxy)benzene, tech
1,2,3-Trimethoxy-5-(prop-2-yn-1-yloxy)benzene, tech
Catalog # APOSOR32363-1G
Supplier:  Apollo Scientific
CAS Number:  
1,2,3-Trimethoxy-5-(prop-2-yn-1-yloxy)benzene, tech
Catalog # APOSOR32363-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :222.237 g/mol
Storage Temperature:Ambient
CAS Number:860787-50-4
Formula:C₁₂H₁₄O₄
MDL Number:MFCD05975060