3-[(4-Chlorophenyl)methyl]-5,5-dimethylimidazolidine-2,4-dione, tech
Katalogové číslo APOSOR32359-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:252.697 g/mol
Storage Temperature:Ambient
CAS Number:860787-34-4
Formula:C₁₂H₁₃ClN₂O₂
MDL Number:MFCD05975050